Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_172595.4 | NP_766183.1 | 5 | 903 | |
GenBank mRNA | AK039965.1 | - | 5 | 867 | |
AK048736.1 | - | 5 | 786 | ||
AK050979.1 | - | 5 | 818 | ||
AK052164.1 | - | 5 | 852 | ||
AK053953.1 | - | 6 | 970 | ||
BC120664.1 | - | 5 | 789 | 40129587 | |
BC120666.1 | - | 5 | 789 | 40129589 | |
BC144993.1 | - | 4 | 729 | 9053541 |