Gene Transcripts

Gene Symbol
CLDN11
Entrez Gene ID
5010
Gene Name
claudin 11
Gene Aliases
OSP, OTM
Chromosome Location
Chr. 3: 170418865 - 170434691 on Build GRCh38
UniGene ID
Hs.31595
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001185056.1 NP_001171985.1 3 600
NM_005602.5 NP_005593.2 3 930
GenBank mRNA AK294087.1 - 3 571
BC013577.1 - 3 918 3895040

View Full Product Details