Gene Transcripts

Gene Symbol
Rtl1
Entrez Gene ID
353326
Gene Name
retrotransposon-like 1
Gene Aliases
6430411K18Rik, Mar, Mart1, Mor1, Peg11
Chromosome Location
Chr. 12: 109590169 - 109595403 on Build GRCm38
UniGene ID
Mm.23062
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_184109.1 NP_908998.1 1 848
GenBank mRNA EU434913.1 - 970
EU434918.1 - 995

View Full Product Details