Gene Transcripts

Gene Symbol
Trpm1
Entrez Gene ID
17364
Gene Name
transient receptor potential cation channel, subfamily M, member 1
Gene Aliases
4732499L03Rik, AI606771, Ltrpc1, Mlsn1
Chromosome Location
Chr. 7: 64153835 - 64269772 on Build GRCm38
UniGene ID
Mm.297430
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK192880.1 - 167

Gene Symbol
Mrgprb1
Entrez Gene ID
233231
Gene Name
MAS-related GPR, member B1
Gene Aliases
MrgB1
Chromosome Location
Chr. 7: 48444113 - 48456342 on Build GRCm38
UniGene ID
Mm.297430
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_205810.4 NP_991379.3 2 945
GenBank mRNA AK148044.1 - 2 941
AK148046.1 - 2 849
AK148052.1 - 2 849
AK148074.1 - 2 946
AK148108.1 - 2 850
AK148114.1 - 2 946
AK148166.1 - 2 850
AK148226.1 - 1 526
AK148241.1 - 2 850
AK148253.1 - 2 941
AK148256.1 - 2 941
AK148273.1 - 2 918
AK148279.1 - 2 899
AK148322.1 - 2 883
AK148388.1 - 2 850
BC116395.1 - 1 771
BC116396.1 - 1 766

View Full Product Details