Gene Transcripts

Gene Symbol
Hoxc13
Entrez Gene ID
15422
Gene Name
homeobox C13
Gene Aliases
Chromosome Location
Chr. 15: 102921116 - 102929135 on Build GRCm38
UniGene ID
Mm.207062
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010464.2 NP_034594.1 2 1151
GenBank mRNA AF193796.1 - 2 1151
AY349616.1 - 2 1179

View Full Product Details