Gene Transcripts

Gene Symbol
Siva1
Entrez Gene ID
30954
Gene Name
SIVA1, apoptosis-inducing factor
Gene Aliases
CD27bp, Siva
Chromosome Location
Chr. 12: 112644828 - 112649152 on Build GRCm38
UniGene ID
Mm.289812
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013929.2 NP_038957.2 1, 2 253
GenBank mRNA AF033113.1 - 1, 2 114
AF033114.1 - 1, 2 201
AK010562.1 - 1, 2 179
AK017570.1 - 1, 2 214
AK153115.1 - 1, 2 163
AK169200.1 - 1, 2 206
AK184852.1 - 1, 2 58
AK209128.1 - 1, 2 58
BC012279.1 - 1, 2 235 4013026
CB524656.1 - 1, 2 402

View Full Product Details