Gene Transcripts

Gene Symbol
CDK7
Entrez Gene ID
1022
Gene Name
cyclin dependent kinase 7
Gene Aliases
CAK, CAK1, CDKN7, HCAK, MO15, STK1, p39MO15
Chromosome Location
Chr. 5: 69234795 - 69277430 on Build GRCh38
UniGene ID
Hs.184298
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001324069.1 NP_001310998.1 11, 12 1140
NM_001324070.1 NP_001310999.1 7, 8 882
NM_001324071.1 NP_001311000.1 8, 9 916
NM_001324072.1 NP_001311001.1 10, 11 917
NM_001324074.1 NP_001311003.1 9, 10 1019
NM_001324075.1 NP_001311004.1 10, 11 1051
NM_001324077.1 NP_001311006.1 10, 11 919
NM_001324078.1 NP_001311007.1 9, 10 1017
NM_001799.3 NP_001790.1 10, 11 1053
NR_136690.1 9, 10 934
XM_011543094.2 XP_011541396.1 7, 8 723
GenBank mRNA AK026509.1 - 10, 11 938
BC000834.2 - 10, 11 892 3455907
BC005298.1 - 10, 11 960 3997991
KU177970.1 - 10, 11 872
KU177971.1 - 9, 10 836
L20320.1 - 11, 12 1047
X77303.1 - 10, 11 915
X77743.1 - 10, 11 906
X79193.1 - 10, 11 962
Y13120.1 - 10, 11 975

View Full Product Details