Gene Transcripts

Gene Symbol
Igflr1
Entrez Gene ID
101883
Gene Name
IGF-like family receptor 1
Gene Aliases
F630003I17Rik, Tmem149
Chromosome Location
Chr. 7: 30565427 - 30567962 on Build GRCm38
UniGene ID
Mm.333757
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145580.2 NP_663555.2 4 717
GenBank mRNA AK154129.1 - 4 718
BC019733.1 - 3 758 3676256

View Full Product Details