Gene Transcripts

Gene Symbol
Iqcj
Entrez Gene ID
208426
Gene Name
IQ motif containing J
Gene Aliases
D230050A05, EG208426
Chromosome Location
Chr. 3: 67892220 - 68056593 on Build GRCm38
UniGene ID
Mm.246568
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177585.3 NP_808253.1 4 970
GenBank mRNA AK052136.1 - 4 971

View Full Product Details