Gene Transcripts

Gene Symbol
Frmpd3
Entrez Gene ID
245643
Gene Name
FERM and PDZ domain containing 3
Gene Aliases
B930041G04, EG245643
Chromosome Location
Chr. X: 140293989 - 140394555 on Build GRCm38
UniGene ID
Mm.208683
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177750.6 NP_808418.1 6 1378
GenBank mRNA AK047241.1 - 4 3098
BC117734.1 - 4 1034 40089821
BC117735.1 - 4 1034 40089822
BC129957.1 - 4 1034 40089827
BC129958.1 - 4 1034 40089829

View Full Product Details