Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001286123.1 | NP_001273052.1 | 2 | 248 | |
NM_001286125.1 | NP_001273054.1 | 2 | 248 | ||
NM_005835.3 | NP_005826.1 | 3 | 698 | ||
XM_005248784.2 | XP_005248841.1 | 3 | 698 | ||
XM_006714949.3 | XP_006715012.1 | 3 | 435 | ||
XM_006714950.2 | XP_006715013.1 | 2 | 325 | ||
XM_006714951.1 | XP_006715014.1 | 3 | 698 | ||
XM_017010159.1 | XP_016865648.1 | 2 | 588 | ||
XM_017010160.1 | XP_016865649.1 | 3 | 693 | ||
GenBank mRNA | BC104822.1 | - | 2 | 221 | |
BC112033.1 | - | 2 | 221 | 8327501 | |
BC143323.1 | - | 2 | 219 | ||
U90544.1 | - | 3 | 583 |