Gene Transcripts

Gene Symbol
Abhd16b
Entrez Gene ID
241850
Gene Name
abhydrolase domain containing 16B
Gene Aliases
1700095H12
Chromosome Location
Chr. 2: 181493206 - 181494980 on Build GRCm38
UniGene ID
Mm.87455
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_183181.2 NP_899004.1 1 694
GenBank mRNA AK028091.1 - 1 696
BC050777.1 - 1 690 6774737

View Full Product Details