Gene Transcripts

Gene Symbol
Cdca7l
Entrez Gene ID
217946
Gene Name
cell division cycle associated 7 like
Gene Aliases
BC006933, JPO2
Chromosome Location
Chr. 12: 117843475 - 117878712 on Build GRCm38
UniGene ID
Mm.281149
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146040.1 NP_666152.1 3 300
GenBank mRNA AK149673.1 - 3 278
BC006933.1 - 3 300 3602150
BC071198.1 - 3 267

View Full Product Details