Gene Transcripts

Gene Symbol
Cep68
Entrez Gene ID
216543
Gene Name
centrosomal protein 68
Gene Aliases
6030463E10Rik, AI481761, BC027174, Kiaa0582
Chromosome Location
Chr. 11: 20226688 - 20250127 on Build GRCm38
UniGene ID
Mm.260314
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172260.3 NP_758464.2 3 772
GenBank mRNA AK031622.1 - 3 769
AK140820.1 - 3 773
AK146669.1 - 3 720
AK162594.1 - 3 778
AK169934.1 - 3 768
AK172975.1 - 3 703
BC027174.1 - 3 766 3154964

View Full Product Details