Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001163847.1 | NP_001157319.1 | 3 | 1175 | |
NM_001163848.1 | NP_001157320.1 | 3 | 1261 | ||
NM_001163849.1 | NP_001157321.1 | 3 | 1175 | ||
NM_001163850.1 | NP_001157322.1 | 3 | 1175 | ||
NM_001163851.1 | NP_001157323.1 | 3 | 1261 | ||
NM_001163852.1 | NP_001157324.1 | 4 | 1411 | ||
NM_001163853.1 | NP_001157325.1 | 3 | 1239 | ||
NM_173186.4 | NP_775278.3 | 3 | 1261 | ||
GenBank mRNA | AK036088.1 | - | 3 | 1128 | |
AK045194.1 | - | 3 | 1147 | ||
AK049306.1 | - | 3 | 1240 | ||
AK049754.1 | - | 3 | 1149 | ||
AK083833.1 | - | 3 | 1147 | ||
AK084693.1 | - | 3 | 1215 | ||
AK086293.1 | - | 3 | 1220 | ||
AK129304.1 | - | 3 | 1193 | ||
AK138431.1 | - | 3 | 1136 | ||
AK147647.1 | - | 3 | 1261 | ||
AK161054.1 | - | 4 | 1413 | ||
BC080845.1 | - | 1 | 1026 | 6850962 | |
BC094417.1 | - | 2 | 1095 | 30943242 |