Gene Transcripts

Gene Symbol
Samd11
Entrez Gene ID
231004
Gene Name
sterile alpha motif domain containing 11
Gene Aliases
BC026991, mr-s
Chromosome Location
Chr. 4: 156246966 - 156255657 on Build GRCm38
UniGene ID
Mm.406833
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001110516.1 NP_001103986.1 9 1415
GenBank mRNA AK149268.1 - 9 1415
AK161035.1 - 8 1429
AY458844.1 - 8 1232
BC026511.1 - 4 1303
BC026991.1 - 8 1547 5363470

View Full Product Details