Gene Transcripts

Gene Symbol
Fam43a
Entrez Gene ID
224093
Gene Name
family with sequence similarity 43, member A
Gene Aliases
Tuf1
Chromosome Location
Chr. 16: 30599723 - 30602797 on Build GRCm38
UniGene ID
Mm.28244
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177632.3 NP_808300.1 1 3012
GenBank mRNA AK083036.1 - 1 3013
BY428362.1 - 1 301

View Full Product Details