Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_182959.3 | NP_892004.1 | 7 | 1220 | |
GenBank mRNA | AF510321.1 | - | 7 | 891 | |
AK082743.1 | - | 7 | 968 | ||
AK149687.1 | - | 5 | 623 | ||
BC042593.1 | - | 7 | 1235 | 5367275 |