Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_013468.3 | NP_038496.2 | 7 | 754 | |
GenBank mRNA | AF041847.1 | - | 7 | 705 | |
AK009655.1 | - | 7 | 747 | ||
AK009959.1 | - | 7 | 753 | ||
AK145944.1 | - | 7 | 795 | ||
AK146471.1 | - | 7 | 751 | ||
AK146627.1 | - | 7 | 753 | ||
AK146940.1 | - | 7 | 751 | ||
BC037138.1 | - | 7 | 754 | 4953125 |