Gene Transcripts

Gene Symbol
Prosc
Entrez Gene ID
114863
Gene Name
proline synthetase co-transcribed
Gene Aliases
1700024N20Rik, 2200002F22Rik
Chromosome Location
Chr. 8: 27042555 - 27056132 on Build GRCm38
UniGene ID
Mm.277501
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039077.2 NP_001034166.1 5 415
NM_001039078.2 NP_001034167.1 4 379
NM_054057.4 NP_473398.1 5 415
GenBank mRNA AK032308.1 - 5 365
AK049905.1 - 4 3548
AK134549.1 - 5 348
AK147670.1 - 5 397
AK160172.1 - 5 369
AK211128.1 - 1 37
AK213501.1 - 2 53
AK218333.1 - 2 53
BC047992.1 - 4 336 6504579
BC061045.1 - 5 409 6534037

View Full Product Details