Gene Transcripts
- Gene Symbol
- RTN3
- Entrez Gene ID
- 10313
- Gene Name
- reticulon 3
- Gene Aliases
- ASYIP, HAP, NSPL2, NSPLII, RTN3-A1
- Chromosome Location
- Chr. 11: 63681315 - 63759891 on Build GRCh38
- UniGene ID
- Hs.743229
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001265589.1 |
NP_001252518.1
|
4, 5 |
2920 |
|
NM_001265590.1 |
NP_001252519.1
|
3, 4 |
2584 |
|
NM_006054.3 |
NP_006045.1
|
2, 3 |
532 |
|
NM_201428.2 |
NP_958831.1
|
3, 4 |
2863 |
|
NM_201429.2 |
NP_958832.1
|
3, 4 |
589 |
|
NM_201430.2 |
NP_958833.1
|
2, 3 |
532 |
|
XM_011544730.2 |
XP_011543032.1
|
4, 5 |
3010 |
|
XM_011544731.2 |
XP_011543033.1
|
3, 4 |
724 |
|
GenBank mRNA |
AB209771.1 |
- |
2, 3 |
469 |
|
AF059524.1 |
- |
2, 3 |
469 |
|
AF119297.1 |
- |
2, 3 |
481 |
|
AK075412.1 |
- |
2, 3 |
482 |
|
AK122727.1 |
- |
2, 3 |
985 |
|
AK127079.1 |
- |
3, 4 |
2559 |
|
AK222898.1 |
- |
2, 3 |
469 |
|
AK295361.1 |
- |
3, 4 |
494 |
|
AK295534.1 |
- |
2, 3 |
386 |
|
AK295758.1 |
- |
3, 4 |
2512 |
|
AK304100.1 |
- |
4, 5 |
2870 |
|
AK307811.1 |
- |
3, 4 |
2809 |
|
AK308083.1 |
- |
2, 3 |
477 |
|
AK309766.1 |
- |
3, 4 |
2538 |
|
AY427821.2 |
- |
3, 4 |
2830 |
|
AY750848.1 |
- |
4, 5 |
2887 |
|
BC000634.1 |
- |
2, 3 |
440 |
|
BC010556.1 |
- |
2, 3 |
445 |
|
BC011394.1 |
- |
2, 3 |
481 |
|
BC022993.1 |
- |
2, 3 |
471 |
|
BC100822.2 |
- |
2, 3 |
462 |
|
BC100823.2 |
- |
2, 3 |
462 |
|
BC105981.1 |
- |
2, 3 |
413 |
|
BC105982.1 |
- |
2, 3 |
444 |
|
BC118550.1 |
- |
2, 3 |
462 |
|
BC118628.1 |
- |
2, 3 |
462 |
|
View Full Product Details