Gene Transcripts

Gene Symbol
Luc7l2
Entrez Gene ID
192196
Gene Name
LUC7-like 2 (S. cerevisiae)
Gene Aliases
4930471C18Rik, AA522013, AU015269, CGI59, Cgi74
Chromosome Location
Chr. 6: 38551339 - 38609471 on Build GRCm38
UniGene ID
Mm.276133
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001170848.1 NP_001164319.1 9 2017
NM_138680.2 NP_619621.2 10 2047
GenBank mRNA AF318301.1 - 10 1972
AK134191.1 - 9 1931
AK147889.1 - 12 2187
AK153527.1 - 13 2275
BC056354.1 - 10 2045 6811460
BC056970.1 - 10 2045 6811460

View Full Product Details