Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_173747.3 | NP_776108.2 | 5 | 812 | |
GenBank mRNA | AK028433.1 | - | 5 | 734 | |
AK144167.1 | - | 5 | 742 | ||
BC062646.1 | - | 3 | 362 | ||
BC092224.1 | - | 5 | 718 | 30355916 | |
BC120679.1 | - | 4 | 495 | 40129602 | |
BC125335.1 | - | 4 | 495 | 40129850 |