Gene Transcripts

Gene Symbol
Gorab
Entrez Gene ID
98376
Gene Name
golgin, RAB6-interacting
Gene Aliases
AI467484, NTKL-BP1, Scyl1bp1
Chromosome Location
Chr. 1: 163384906 - 163403908 on Build GRCm38
UniGene ID
Mm.32901
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178883.5 NP_849214.2 2 177
GenBank mRNA AK039170.1 - 2 184
AK043953.1 - 2 178
AK165909.1 - 2 147
AK169151.1 - 2 170
AK170455.1 - 2 194
BC119132.1 - 2 160 8733881
BC119134.1 - 2 160 8733883

View Full Product Details