Gene Transcripts

Gene Symbol
Rftn1
Entrez Gene ID
76438
Gene Name
raftlin lipid raft linker 1
Gene Aliases
2310015N21Rik, AI853526
Chromosome Location
Chr. 17: 49993258 - 50190497 on Build GRCm38
UniGene ID
Mm.41854
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_181397.2 NP_852062.1 2 386
GenBank mRNA AK087803.1 - 159
AK152130.1 - 2 308
AK153288.1 - 2 308
AK155870.1 - 2 350
AK170913.1 - 2 350
AK172884.1 - 2 314
BC052074.1 - 2 386 6401192

View Full Product Details