Gene Transcripts

Gene Symbol
Spred3
Entrez Gene ID
101809
Gene Name
sprouty-related, EVH1 domain containing 3
Gene Aliases
AW047143, D130060H24Rik, Spred-3
Chromosome Location
Chr. 7: 29157783 - 29169854 on Build GRCm38
UniGene ID
Mm.377262
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_182927.3 NP_891557.3 6 3324
GenBank mRNA AK051617.1 - 3 2826
BC062119.1 - 6 3323 6821036

View Full Product Details