Gene Transcripts

Gene Symbol
Ecd
Entrez Gene ID
70601
Gene Name
ecdysoneless homolog (Drosophila)
Gene Aliases
5730461K03Rik, Hsgt1
Chromosome Location
Chr. 14: 20319859 - 20348121 on Build GRCm38
UniGene ID
Mm.35105
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027475.3 NP_081751.1 5 850
GenBank mRNA AK017681.1 - 3 274
AK028664.1 - 5 814
AK029169.1 - 5 987
AK043261.1 - 5 699
BC013470.1 - 5 799 4220504

View Full Product Details