Gene Transcripts
- Gene Symbol
- Cic
- Entrez Gene ID
- 71722
- Gene Name
- capicua homolog (Drosophila)
- Gene Aliases
- 1200010B10Rik, mKIAA0306
- Chromosome Location
- Chr. 7: 25267657 - 25294159 on Build GRCm38
- UniGene ID
- Mm.28833
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AF363690.1 |
- |
3 |
376 |
|
AK147276.1 |
- |
3 |
1009 |
|
AK147535.1 |
- |
4 |
3127 |
|
View Full Product Details