Gene Transcripts

Gene Symbol
CNKSR1
Entrez Gene ID
10256
Gene Name
connector enhancer of kinase suppressor of Ras 1
Gene Aliases
CNK, CNK1
Chromosome Location
Chr. 1: 26177490 - 26189884 on Build GRCh38
UniGene ID
Hs.16232
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001297647.1 NP_001284576.1 3 284
NM_001297648.1 NP_001284577.1 3 284
NM_006314.2 NP_006305.2 3 284
XM_011540484.2 XP_011538786.1 3 264
GenBank mRNA AB209335.1 - 3 279
AF100153.1 - 3 262
AK001900.1 - 3 263
AK222817.1 - 3 263
AK222904.1 - 3 267
AK292646.1 - 3 266
AK302056.1 - 3 265
BC011604.2 - 3 265 3139227
BC012797.2 - 3 265 3505293
BT006900.1 - 3 226
DA868484.1 - 3 265
DR159101.1 - 3 227

View Full Product Details