Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_028447.3 | NP_082723.2 | 7 | 1142 | |
GenBank mRNA | AK004443.1 | - | 7 | 1142 | |
AK143537.1 | - | 7 | 1134 | ||
AK147610.1 | - | 1 | 875 | ||
BC014817.1 | - | 7 | 1058 | 3709357 |