Gene Transcripts

Gene Symbol
Lmbr1l
Entrez Gene ID
74775
Gene Name
limb region 1 like
Gene Aliases
1110013E13Rik, AA415653, D15Ertd735e, Limr
Chromosome Location
Chr. 15: 98903914 - 98918550 on Build GRCm38
UniGene ID
Mm.385020
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029098.3 NP_083374.1 4 474
GenBank mRNA AK003656.1 - 4 406
AK030589.1 - 4 440
AK049110.1 - 4 475
AK143891.1 - 4 435
AK154968.1 - 4 608
AK170328.1 - 4 435
AK173121.1 - 4 855
AY052398.1 - 4 317
BC023397.1 - 4 411 5043877

View Full Product Details