Gene Transcripts

Gene Symbol
Vmp1
Entrez Gene ID
75909
Gene Name
vacuole membrane protein 1
Gene Aliases
3110098I04Rik, 4930579A11Rik, AI787464, Tango5, Tmem49, mir-21a, ni-2
Chromosome Location
Chr. 11: 86583852 - 86683822 on Build GRCm38
UniGene ID
Mm.390398
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029478.3 NP_083754.2 12 2019
GenBank mRNA AK043091.1 - 12 2020
AK049559.1 - 12 2012

View Full Product Details