Gene Transcripts

Gene Symbol
Hspa12a
Entrez Gene ID
73442
Gene Name
heat shock protein 12A
Gene Aliases
1700063D12Rik, AI118035, AI840429, AW048913, AW556406, D5Mgi40, mKIAA0417
Chromosome Location
Chr. 19: 58795750 - 58943545 on Build GRCm38
UniGene ID
Mm.39739
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175199.3 NP_780408.1 8 943
GenBank mRNA AB093239.1 - 8 887
AK038491.1 - 8 915
AK087732.1 - 9 1101
AK141925.1 - 8 944
AY196789.1 - 8 942
BC030362.1 - 8 913 5369051

View Full Product Details