Gene Transcripts
- Gene Symbol
- AKAP3
- Entrez Gene ID
- 10566
- Gene Name
- A-kinase anchoring protein 3
- Gene Aliases
- AKAP 110, AKAP110, CT82, FSP95, HEL159, PRKA3, SOB1
- Chromosome Location
- Chr. 12: 4615508 - 4649047 on Build GRCh38
- UniGene ID
- Hs.98397
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001278309.1 |
NP_001265238.1
|
4, 5 |
639 |
|
NM_006422.3 |
NP_006413.3
|
4, 5 |
617 |
|
XM_005253664.2 |
XP_005253721.1
|
4, 5 |
549 |
|
XM_011520908.1 |
XP_011519210.1
|
3, 4 |
753 |
|
XM_011520909.1 |
XP_011519211.1
|
2, 3 |
230 |
|
XM_011520910.2 |
XP_011519212.1
|
2, 3 |
685 |
|
GenBank mRNA |
AF087003.1 |
- |
3, 4 |
254 |
|
AF093408.1 |
- |
3, 4 |
321 |
|
AK292451.1 |
- |
4, 5 |
617 |
|
BC047535.1 |
- |
3, 4 |
301 |
5267552 |
EU794681.1 |
- |
3, 4 |
321 |
|
U85715.1 |
- |
3, 4 |
294 |
|
View Full Product Details