Gene Transcripts

Gene Symbol
SMO
Entrez Gene ID
6608
Gene Name
smoothened, frizzled class receptor
Gene Aliases
FZD11, Gx, SMOH
Chromosome Location
Chr. 7: 129188872 - 129213548 on Build GRCh38
UniGene ID
Hs.437846
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005631.4 NP_005622.1 3 831
XM_011516522.2 XP_011514824.1 4 397
GenBank mRNA AF071494.1 - 3 551
AF120103.1 - 3 609
BC009989.2 - 3 831 4127774
U84401.1 - 3 551

View Full Product Details