Gene Transcripts

Gene Symbol
TGOLN2
Entrez Gene ID
10618
Gene Name
trans-golgi network protein 2
Gene Aliases
TGN38, TGN46, TGN48, TGN51, TTGN2
Chromosome Location
Chr. 2: 85318018 - 85328296 on Build GRCh38
UniGene ID
Hs.593382
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001206840.1 NP_001193769.1 2 855
NM_001206841.1 NP_001193770.1 2 855
NM_001206844.1 NP_001193773.1 2 855
NM_006464.3 NP_006455.2 2 855
GenBank mRNA AF027515.1 - 2 583
AF027516.1 - 2 583
AK126465.1 - 3 252
AK222829.1 - 2 547
AK223063.1 - 2 546
AK312479.1 - 2 548
BC008461.1 - 2 548 4272702
BC028219.1 - 2 544 5217823
BX640868.1 - 2 811
U62390.1 - 2 583
X94333.1 - 2 539

View Full Product Details