Gene Transcripts

Gene Symbol
Ube2r2
Entrez Gene ID
67615
Gene Name
ubiquitin-conjugating enzyme E2R 2
Gene Aliases
1200003M11Rik, Cdc34b, Ubc3b
Chromosome Location
Chr. 4: 41136021 - 41193370 on Build GRCm38
UniGene ID
Mm.389540
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026275.4 NP_080551.1 5 2799
GenBank mRNA AK075714.1 - 5 2801

View Full Product Details