Gene Transcripts

Gene Symbol
Bend5
Entrez Gene ID
67621
Gene Name
BEN domain containing 5
Gene Aliases
2310026E23Rik
Chromosome Location
Chr. 4: 111414540 - 111460298 on Build GRCm38
UniGene ID
Mm.45533
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026279.3 NP_080555.1 3 565
GenBank mRNA AK014057.1 - 3 555
AK075885.1 - 3 565
AK156708.1 - 3 467
AK167425.1 - 3 385
BC037633.1 - 3 398

View Full Product Details