Gene Transcripts

Gene Symbol
Rabl3
Entrez Gene ID
67657
Gene Name
RAB, member RAS oncogene family-like 3
Gene Aliases
4930553C05Rik, AA958834
Chromosome Location
Chr. 16: 37539858 - 37572385 on Build GRCm38
UniGene ID
Mm.28158
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001042499.1 NP_001035964.1 8 969
GenBank mRNA AK011280.1 - 8 911
AK016099.1 - 8 978
AK028195.1 - 8 963
AK028269.1 - 8 956
BC050194.1 - 8 950 30022531

View Full Product Details