Gene Transcripts

Gene Symbol
Lipf
Entrez Gene ID
67717
Gene Name
lipase, gastric
Gene Aliases
2310051B21Rik, AV082900
Chromosome Location
Chr. 19: 33919441 - 33976813 on Build GRCm38
UniGene ID
Mm.329816
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026334.3 NP_080610.1 5 513
GenBank mRNA AK009300.1 - 5 515
AK009359.1 - 5 515
AK009413.1 - 5 514
AK009428.1 - 5 517
AK009431.1 - 5 517
AK009437.1 - 5 515
AK009459.1 - 5 515
AK009473.1 - 5 514
AK009474.1 - 5 515
AK009479.1 - 5 512
AK009523.1 - 5 515
AK009525.1 - 5 515
AK009537.1 - 5 515
AK009544.1 - 5 510
AK009546.1 - 5 518
AK009560.1 - 5 515
AK009571.1 - 5 514
AK009573.1 - 5 518
AK009729.1 - 5 515
AK009773.1 - 5 515
AK009875.1 - 5 515
AK009923.1 - 5 577
AK009990.1 - 5 515
AK010019.1 - 5 516
AK010026.1 - 5 515
AK010035.1 - 5 515
AK010036.1 - 5 513
AK010058.1 - 5 514
AK010061.1 - 5 515
AK010091.1 - 5 513
AK010093.1 - 5 515
AK010103.1 - 5 516
AK010106.1 - 5 515
AK010116.1 - 5 513
AK010123.1 - 5 516
AK010124.1 - 4 509
AK010125.1 - 5 515
AK010139.1 - 5 515
AK010148.1 - 5 515
AK010173.1 - 5 515
AK010203.1 - 5 516
AK010231.1 - 5 514
AK010236.1 - 5 516
AK075913.1 - 5 518
BC061067.1 - 5 513 30301084

View Full Product Details