Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_027346.1 | NP_081622.1 | 1 | 370 | |
GenBank mRNA | AK010072.1 | - | 1 | 388 | |
AK034014.1 | - | 1 | 417 | ||
AK047078.1 | - | 1 | 390 | ||
AK140143.1 | - | 1 | 408 | ||
AK147942.1 | - | 1 | 406 | ||
AK167963.1 | - | 1 | 387 | ||
BC029182.1 | - | 1 | 370 | 4195338 |