Gene Transcripts

Gene Symbol
Fam25c
Entrez Gene ID
69134
Gene Name
family with sequence similarity 25, member C
Gene Aliases
2200001I15Rik, Fam25
Chromosome Location
Chr. 14: 34351882 - 34355393 on Build GRCm38
UniGene ID
Mm.27156
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_183278.2 NP_899101.1 3 347
GenBank mRNA AK008614.1 - 3 347
BC086894.1 - 3 349

View Full Product Details