Gene Transcripts

Gene Symbol
Nsa2
Entrez Gene ID
59050
Gene Name
NSA2 ribosome biogenesis homolog (S. cerevisiae)
Gene Aliases
5730427N09Rik, LNR42, Nsa2p, Tinp1, Yr29
Chromosome Location
Chr. 13: 97129427 - 97137926 on Build GRCm38
UniGene ID
Mm.390634
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021552.5 NP_067527.3 4, 5 683
GenBank mRNA AF238866.1 - 4, 5 609
AK002474.1 - 3, 4 534
AK009136.1 - 3, 4 612
AK012832.1 - 4, 5 624
AK014275.1 - 4, 5 660
AK017605.1 - 4, 5 624
AK078457.1 - 4, 5 636
BC086776.1 - 3, 4 479
BC116384.1 - 4, 5 624
BC126976.1 - 4, 5 623 40101726
BC172160.1 - 4, 5 595

View Full Product Details