Gene Transcripts

Gene Symbol
Tmem42
Entrez Gene ID
66079
Gene Name
transmembrane protein 42
Gene Aliases
0610027O18Rik, 4933429E06Rik, AV003444, D9Ertd662e
Chromosome Location
Chr. 9: 123021038 - 123023491 on Build GRCm38
UniGene ID
Mm.482265
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025339.4 NP_079615.1 3 733
GenBank mRNA AK002688.1 - 3 709
AK012820.1 - 3 708
BC049583.1 - 3 733 6742203

View Full Product Details