Gene Transcripts

Gene Symbol
Prr13
Entrez Gene ID
66151
Gene Name
proline rich 13
Gene Aliases
1110020C13Rik, 2010324E22Rik
Chromosome Location
Chr. 15: 102459170 - 102462806 on Build GRCm38
UniGene ID
Mm.393955
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001170911.1 NP_001164382.1 4 707
NM_025385.3 NP_079661.1 5 825
GenBank mRNA AK002430.1 - 4 675
AK003850.1 - 5 822
AK008611.1 - 3 647
AK008894.1 - 5 785
AK154583.1 - 5 785
BC016234.1 - 4 971 4503179

View Full Product Details