Gene Transcripts

Gene Symbol
Timm50
Entrez Gene ID
66525
Gene Name
translocase of inner mitochondrial membrane 50
Gene Aliases
2810403L02Rik, AU015082, TIM50L
Chromosome Location
Chr. 7: 28305516 - 28312046 on Build GRCm38
UniGene ID
Mm.167913
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025616.3 NP_079892.1 9, 10 876
GenBank mRNA AK008340.1 - 9, 10 877
AK008382.1 - 9, 10 873
AK010832.1 - 9, 10 842
AK088074.1 - 9, 10 867
AK167473.1 - 9, 10 879
AY551342.1 - 9, 10 862
BC010303.1 - 9, 10 851 3597676
BC025844.1 - 3, 4 357
BC037056.1 - 9, 10 878

View Full Product Details