Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_025808.3 | NP_080084.2 | 1 | 195 | |
GenBank mRNA | AK004561.2 | - | 1 | 195 | |
AK005037.1 | - | 1 | 194 | ||
AK083411.1 | - | 1 | 199 | ||
AK132817.1 | - | 1 | 194 | ||
BC034400.1 | - | 1 | 192 | 2651456 |