Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_025891.3 | NP_080167.3 | 5 | 614 | |
GenBank mRNA | AK005094.1 | - | 5 | 561 | |
AK158019.1 | - | 589 | |||
BC013122.1 | - | 5 | 651 | 3586276 | |
BC060525.1 | - | 5 | 513 | 6817219 |