Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_016909.2 | NP_058605.1 | 3 | 358 | |
GenBank mRNA | AF187040.1 | - | 3 | 358 | |
AK014437.1 | - | 3 | 306 | ||
AK030407.1 | - | 3 | 288 | ||
AK146339.1 | - | 3 | 291 | ||
AK159223.1 | - | 3 | 360 | ||
AK164453.1 | - | 2 | 133 | ||
AK189358.1 | - | 3 | 163 | ||
BC004611.1 | - | 3 | 260 | 3585400 |