Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001290745.1 | NP_001277674.1 | 11 | 1201 | |
NM_016965.3 | NP_058661.1 | 10 | 1183 | ||
GenBank mRNA | AJ534525.1 | - | 10 | 966 | |
AK122318.1 | - | 9 | 1004 | ||
BC060197.1 | - | 10 | 1162 | ||
BC127067.1 | - | 11 | 983 | ||
X61453.1 | - | 2 | 105 |